Imprimer
Hedland Flowmeter Accessories
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
03231-80 03231-85
MFLX03231-80EA 201 EUR
MFLX03231-80 MFLX03231-85
Hedland Flowmeter Accessories
Flow Switches
Make the most of your Hedland flowmeters with these replacement parts and other accessories.
  • Optimize performance of your flowmeter with these accessories
Replacement switches and cables are suitable for use with Hedland spring-loaded in-line flowmeters or turbine flowmeters.