Imprimer
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, Polypropylene, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX40610-72EA 29.9 EUR
MFLX40610-72 MFLX40610-73 MFLX40610-74 MFLX40610-77 MFLX40610-82 MFLX40610-83 MFLX40624-09 MFLX40640-73 MFLX40610-75 MFLX40610-76 MFLX40610-78 MFLX40610-79 MFLX40610-85 MFLX40610-86 MFLX40610-87 MFLX40610-88 MFLX40640-75 MFLX40640-78 MFLX40640-81
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, Polypropylene, Avantor®
Raccords et connecteurs
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.