Imprimer
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, HDPE, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30624-00EA 24.1 EUR
MFLX30624-00 MFLX30624-06 MFLX30624-10 MFLX30624-16 MFLX30624-20 MFLX30624-24 MFLX30401-65 MFLX30401-67 MFLX30623-86 MFLX30623-88 MFLX30623-96 MFLX30624-04 MFLX30624-12 MFLX30624-18 MFLX30624-22 MFLX30627-84 MFLX30628-04 MFLX30628-10
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, HDPE, Avantor®
Raccords et connecteurs
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.