Imprimer
Masterflex® Union Fittings, Hose Barb, Tee, PVDF, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40615-60
MFLX40615-60EA 51.7 EUR
MFLX40615-60 MFLX40615-54 MFLX40615-58 MFLX40615-55 MFLX40615-57 MFLX40615-59 MFLX40615-61 MFLX40615-63 MFLX40615-64 MFLX40615-65
Masterflex® Union Fittings, Hose Barb, Tee, PVDF, Avantor®
Raccords et connecteurs
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).