Imprimer
Masterflex® Union Fittings, Hose Barb, Tee, Polypropylene, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30613-51EA 36 EUR
MFLX30613-51 MFLX30613-63 MFLX30613-71 MFLX30613-75 MFLX40610-30 MFLX40610-31 MFLX40610-32 MFLX40610-33 MFLX40610-35 MFLX40623-70 MFLX40623-76 MFLX40627-70 MFLX40627-76 MFLX40640-30 MFLX40640-31 MFLX40640-32 MFLX06365-77 MFLX40613-43 MFLX40613-47 MFLX40613-48 MFLX50623-57 MFLX50623-66 MFLX50624-28 MFLX50624-30 MFLX50627-57 MFLX50627-66 MFLX50628-29 MFLX40640-33 MFLX40640-34
Masterflex® Union Fittings, Hose Barb, Tee, Polypropylene, Avantor®
Raccords et connecteurs
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).