Imprimer
Masterflex® Union Fittings, Hose Barb, Elbow, HDPE, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30622-65EA 30.3 EUR
MFLX30622-65 MFLX30622-67 MFLX30622-69 MFLX30622-72 MFLX30622-76 MFLX30622-78 MFLX30622-80 MFLX30626-76 MFLX40611-95 MFLX30626-72 MFLX40622-63 MFLX40626-63 MFLX41518-47
Masterflex® Union Fittings, Hose Barb, Elbow, HDPE, Avantor®
Raccords et connecteurs
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!