Imprimer
Masterflex® Sanitary Clamp Fittings (Nylon), Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30508-21EA 31.6 EUR
MFLX30508-21 MFLX30508-23 MFLX30508-25 MFLX30508-27 MFLX30508-29
Masterflex® Sanitary Clamp Fittings (Nylon), Avantor®
Raccords et connecteurs
Compatible with metal and plastic Tri-Clamp® fittings.
  • Excellent chemical resistance; low leachable characteristics
  • Autoclavable
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.