Imprimer
Masterflex® Reducer Fittings, Hose Barb, Straight (HDPE), Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30616-33EA 38.4 EUR
MFLX30616-33 MFLX30621-96 MFLX30621-98 MFLX30622-00 MFLX30622-03 MFLX30622-07 MFLX30622-09 MFLX30622-11 MFLX30622-13 MFLX30622-15 MFLX30622-24 MFLX30622-27 MFLX30622-29 MFLX30622-33 MFLX30622-35 MFLX30622-36 MFLX30622-38 MFLX30622-40 MFLX30622-42 MFLX30626-27 MFLX30626-29 MFLX30626-35 MFLX30626-36 MFLX30626-38 MFLX30626-42 MFLX31518-33 MFLX41518-22 MFLX41518-29 MFLX41518-31 MFLX30616-29 MFLX30622-05 MFLX30622-17 MFLX30625-96 MFLX30626-05 MFLX30626-07 MFLX30626-13 MFLX30626-17 MFLX30626-24 MFLX30626-33 MFLX30626-40
Masterflex® Reducer Fittings, Hose Barb, Straight (HDPE), Avantor®
Raccords et connecteurs
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range −65 to +150 °F (−53 to +65 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.