Imprimer
Barblock Fitting and Tubing Tools and Accessories
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30604-12EA 295 EUR
MFLX30604-12 MFLX30604-14 MFLX30604-16 MFLX30604-18
Barblock Fitting and Tubing Tools and Accessories
Outils d'extraction et d'insertion
Easily install your Barblock Fittings.
  • Optimize performance of your Barblock fittings
Select a hand tool to assemble your BarbLock Retainers based on the tubing size. Universal removal tool is also available.