Imprimer
Abaque™ Lubricant for Positive Displacement Pumps
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX75810-89EA 103 EUR
MFLX75810-89 MFLX75810-88
Abaque™ Lubricant for Positive Displacement Pumps
Pompe Pompes péristaltiques
Replacement lubricant for the liquid end coming in four different volumnes.
  • Original lubricant designed to help extend tubing life and to dissipate heat build up.
Optimize performance of your Abaque™ positive displacement pumps.