Imprimer
Masterflex® Ismatec® Microbore Pump Tubing, Tygon® LMT-55, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX05502-10EA 99.3 EUR
MFLX05502-10 MFLX05502-12 MFLX05502-14 MFLX05502-16 MFLX05502-18 MFLX05502-19 MFLX05502-22 MFLX05502-24 MFLX05502-26 MFLX05502-27 MFLX05502-28 MFLX05502-30 MFLX05502-31 MFLX05502-32 MFLX05502-34 MFLX05502-36 MFLX05502-38 MFLX05502-39 MFLX05502-40 MFLX05502-42 MFLX05502-44 MFLX05502-46 MFLX05502-48 MFLX05502-49 MFLX05502-09
Masterflex® Ismatec® Microbore Pump Tubing, Tygon® LMT-55, Avantor®
Tuyaux Tubes pour pompe péristaltique
Crystal clear and flexible while also being long-lasting and crack-resistant.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Designed for optimal operation in your Ismatec pumps.
Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments
Sterilize by autoclave or ethylene oxide gas.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.