Imprimer
Masterflex® L/S® MiniFlex® OEM Pump Heads, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX77220-00EA 288 EUR
MFLX77220-00 MFLX77220-10
Masterflex® L/S® MiniFlex® OEM Pump Heads, Avantor®
Pompe Pump Heads Peristaltic Pump Heads
Pump head accepts continuous lengths of tubing for clean flow paths.
  • Flow range of 1.8 to 220 ml/min—flow rate depends on drove rpm, pump head, and tubing size
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Dual-channel model delivers synchronous flow from two separate channels; single-channel model delivers highest flow rate
  • Automatic tubing retention makes tube loading quick and easy
The Masterflex® Miniflex® pump heads feature a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-En savoir plus