Imprimer
Cadence Science Nickel-Plated Brass Luer Stopcocks
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
41507-00 41507-01 41507-02 41507-10 41507-11 41507-12 41507-13
MFLX41507-00EA 105 EUR
MFLX41507-00 MFLX41507-01 MFLX41507-02 MFLX41507-10 MFLX41507-11 MFLX41507-12 MFLX41507-13
Cadence Science Nickel-Plated Brass Luer Stopcocks
Vannes Ball Valves
Exceptional durability.
  • Easy to disassemble for cleaning
  • Stopcock handles turn 180°
  • Autoclavable
These Cadence Science luer stopcocks are constructed of nickel-plated brass for enhanced durability. These stopcocks offer worry-free cleaning with easy disassembly to save valuable time.