Imprimer
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX07624-18EA 119 EUR
MFLX07624-18 MFLX07624-22 MFLX07624-24 MFLX07624-26 MFLX07624-28 MFLX07624-30 MFLX07624-32 MFLX07624-34 MFLX07624-36 MFLX07624-38 MFLX07624-40 MFLX07624-42 MFLX07624-44 MFLX07624-46 MFLX07624-48
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Tuyaux Tubes pour pompe péristaltique
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attachedEn savoir plus