Imprimer
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-Lab, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX96461-09EA 111 EUR
MFLX96461-09 MFLX96461-10 MFLX96461-12 MFLX96461-14 MFLX96461-16 MFLX96461-18 MFLX96461-19 MFLX96461-22 MFLX96461-24 MFLX96461-26 MFLX96461-27 MFLX96461-28 MFLX96461-29 MFLX96461-30 MFLX96461-31 MFLX96461-32 MFLX96461-34 MFLX96461-36 MFLX96461-38 MFLX96461-39 MFLX96461-40 MFLX96461-42 MFLX96461-44 MFLX96461-46 MFLX96461-48 MFLX96461-49
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-Lab, Avantor®
Tuyaux Tubes pour pompe péristaltique
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Ideal for general laboratory applications
  • Clear for easy sample viewing
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
The Tygon® S3™ E-Lab features an ultra-smooth inner bore, excellent biocompatibility for sensitive applications, non-cytotoxic, non-pyrogenic, and non-hemolytic, low protein binding, heat-sealable, bondable, sterile-weldable, and custom moldable for assemblies. It is ideal for pharmaceutical, biotech, and general-purpose lab applications, including sterile filling and processing, cell harvest and media process systems, filtration, fermentation, cell and tissue culture transport.It features a clear appearance, has a temperature range of –66 to 135 °C, and has a pumping life of approximately 1000 hours at 100 rpm.
The overall length of E-Lab (E-3603) 96461-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.