Imprimer
Masterflex® Ismatec® Microbore Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX06449-10EA 241 EUR
MFLX06449-10 MFLX06449-12 MFLX06449-14 MFLX06449-18 MFLX06449-22 MFLX06449-24 MFLX06449-26 MFLX06449-28 MFLX06449-30 MFLX06449-34 MFLX06449-36 MFLX06449-40 MFLX06449-42 MFLX06449-48 MFLX06449-31 MFLX06449-33 MFLX06449-37 MFLX06449-47
Masterflex® Ismatec® Microbore Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Tuyaux Tubes pour pompe péristaltique
Longest life of all Tygon® formulations.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Applications: General laboratory use
Characteristics: Long life, won’t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.