Imprimer
Masterflex® Ismatec® Microbore Pump Tubing, Puri-Flex®, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX96418-10EA 86.7 EUR
MFLX96418-10 MFLX96418-12 MFLX96418-14 MFLX96418-16 MFLX96418-18 MFLX96418-22 MFLX96418-24 MFLX96418-26 MFLX96418-28 MFLX96418-30 MFLX96418-34 MFLX96418-36 MFLX96418-40 MFLX96418-42 MFLX96418-44 MFLX96418-46 MFLX96418-48 MFLX96418-32
Masterflex® Ismatec® Microbore Pump Tubing, Puri-Flex®, Avantor®
Tuyaux Tubes pour pompe péristaltique
Long life tubing for pharma and bioprocess applications.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Applications: General-purpose lab use requiring high performance and long life
Characteristics: Exceptional tensile and tear strength, good biocompatibility, low gas permeability, smooth surface, translucent
Certifications: FDA-compliant (21 CFR 177.2600), meets USP Class VI requirements, manufactured according to GMP
Temperature range: –58 to 275 °F (–50 to 135 °C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.