Imprimer
Masterflex® Pump Handheld Remote Controllers, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX07523-97EA 485 EUR
MFLX07523-97 MFLX07528-80 MFLX07595-62
Masterflex® Pump Handheld Remote Controllers, Avantor®
Pompe Pompes péristaltiques
Easily control your Masterflex® L/S® pumps while filling and dispensing.
  • Suitable for use with Masterflex® L/S® pumps
  • Long 10ft cable makes remote control simple
These ergonomic handheld controllers are designed to control basic pump drive operations including start/stop, direction, and prime — depending on available drive functions. Route tubing through the handle for convenience in filling and dispensing applications. These handheld controllers accept Masterflex tubing sizes L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18, L/S 15, and L/S 24 (up to 7/16" nominal OD).
Informations de livraison: Includes 6-ft (1.8 m) cable with noted terminal connector.