Imprimer
Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
33402-04 33402-10 33402-12 33402-14 33402-34 33402-38 33402-40 33402-44 33402-48
MFLX33402-04EA 787 EUR
MFLX33402-04 MFLX33402-10 MFLX33402-12 MFLX33402-14 MFLX33402-34 MFLX33402-38 MFLX33402-40 MFLX33402-44 MFLX33402-48
Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms, Avantor®
Débitmètres
Protect equipment with automatic flow rate monitoring via high and low alarms.
  • Protect your equipment from damage caused by extreme flow rates
  • Features dual-mounted LED receiver/sensors with optical sensors
These 150-mm valved flowmeters feature two noninvasive optical sensors that easily glide up and down both sides of the meter to set high and low flow rates. The meter is ideal for use with an alarm, valve, or other devices to control flow with use of NPN and PNP transistor outputs. The compact design and ease of operation make this meter perfect for laboratory and industrial uses.