Imprimer
Masterflex® Reducer Fittings, Hose Barb, Y-Connector, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30726-51EA 46.6 EUR
MFLX30726-51 MFLX30726-52 MFLX30726-53 MFLX30726-54 MFLX30726-55 MFLX30726-56 MFLX30726-57 MFLX30726-11 MFLX30726-12 MFLX30726-13 MFLX30726-14 MFLX30726-16 MFLX30726-17 MFLX30726-31 MFLX30726-32 MFLX30726-34 MFLX30726-35 MFLX30726-36 MFLX30726-37 MFLX30726-33 MFLX40614-93 MFLX40614-94 MFLX40614-95 MFLX40614-96 MFLX40614-97 MFLX40614-98 MFLX40614-99
Masterflex® Reducer Fittings, Hose Barb, Y-Connector, Avantor®
Raccords et connecteurs
Ensure a tight seal every time.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).