Imprimer
Masterflex® Flanged Sanitary Gaskets, Tri-Clamp®, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX30564-28EA 80.9 EUR
MFLX30564-28 MFLX30564-44 MFLX30564-46
Masterflex® Flanged Sanitary Gaskets, Tri-Clamp®, Avantor®
Scellés Joints
Hands-free installation for difficult-to-reach locations!
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
With the same benefits as standard sanitary gaskets, these gaskets offer an outer lip to grip the sanitary coupling port - once in place, no need to worry about maintaining the gasket's placement! An especially useful feature for hard-to-reach or vertical clamp joints, using the outer lip speeds assembly and reduces your line's down time.
Available in platinum-cured silicone, EPDM, Buna N and FKM (see material descriptions under standard sanitary fittings for more information).