Imprimer
CPC® Quick-Disconnect Fittings, Hose Barb Inserts
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06361-71 06364-38 06364-39 06364-42 06361-71BLK 06364-38BLK 06364-39BLK 06364-42BLK 31307-09BLK 31307-14BLK 31307-61BLK 31307-64BLK 31307-69BLK
MFLX06360-36EA 16.3 EUR
MFLX06360-36 MFLX06360-38 MFLX06360-40 MFLX06360-42 MFLX06360-43 MFLX06360-44 MFLX06360-45 MFLX06360-80 MFLX06360-82 MFLX06360-83 MFLX06360-85 MFLX06360-87 MFLX06361-34 MFLX06364-40 MFLX06364-45 MFLX06364-47 MFLX06364-80 MFLX06364-82 MFLX06364-85 MFLX06364-87 MFLX06361-71 MFLX06364-38 MFLX06364-39 MFLX06364-42 MFLX06361-71BLK MFLX06364-38BLK MFLX06364-39BLK MFLX06364-42BLK MFLX06361-34BLK MFLX06364-80BLK MFLX06360-36BLK MFLX06360-40BLK MFLX06360-44BLK MFLX06360-45BLK MFLX06360-82BLK MFLX06364-82BLK MFLX06364-85BLK MFLX06364-40BLK MFLX06364-47BLK MFLX06360-43BLK MFLX06364-45BLK MFLX06360-38BLK MFLX06360-42BLK MFLX06360-87BLK MFLX06360-80BLK MFLX06360-83BLK MFLX06360-85BLK MFLX31307-09BLK MFLX31307-14BLK MFLX31307-61BLK MFLX31307-64BLK MFLX31307-69BLK
CPC® Quick-Disconnect Fittings, Hose Barb Inserts
Raccords et connecteurs
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).