Imprimer
Valved Acrylic Flowmeters for Bench or Panel Mount
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
32470-11 32470-13 32470-18 32470-20 32470-24 32470-37 32470-38 32470-51 32470-52 32470-55
MFLX32470-11EA 134 EUR
MFLX32470-11 MFLX32470-13 MFLX32470-18 MFLX32470-20 MFLX32470-24 MFLX32470-37 MFLX32470-38 MFLX32470-51 MFLX32470-52 MFLX32470-55
Valved Acrylic Flowmeters for Bench or Panel Mount
Débitmètres
Durable single-piece body withstands severe shock and vibration.
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters' inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (32462-50 sold separately) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialits for quotes on acrylic meters with special requirements.