Imprimer
Flexelene™ SFX Soft Flexible Polyethylene Tubing
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
96180-01 96180-02 96180-03 96180-04 96180-05 96180-07
MFLX96180-01EA 134 EUR
MFLX96180-01 MFLX96180-02 MFLX96180-03 MFLX96180-04 MFLX96180-05 MFLX96180-07
Flexelene™ SFX Soft Flexible Polyethylene Tubing
Tuyaux Transfer Tubing
Designed to produce low extractables.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotech, and general lab applications.
Characteristics: Good chemical resistance. Ultra-low extractables. Heat-sealable. Translucent.
Certifications: Meets USP Class VI requirements.
Temperature range: -40 to 100 °F (-40 to 37 °C)
Sterilize: Ethylene oxide or gamma irradiation.