Imprimer
Hedland High TempFlow Meters for Petroleum and Water
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
32508-92 32509-07
MFLX32508-92EA 535 EUR
MFLX32508-92 MFLX32509-07
Hedland High TempFlow Meters for Petroleum and Water
Débitmètres
Direct read 360° rotatable scale.
  • Easy-to-read linear scale
FEATURES
Direct readings
Install in any position
360° Rotatable Guard/Scale
No flow Straighteners or Special Piping Required
Insensitive to Shock and Vibration
Good Viscosity Stability
Temperature up to 500 °F
Accuracy +/- 2% Full Scale
Repeatability +/-1%
Special Scales Available