Imprimer
Masterflex® Turbine Flowmeter Rate Sensors, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
32500-34
MFLX32500-27EA 254 EUR
MFLX32500-27 MFLX32500-34
Masterflex® Turbine Flowmeter Rate Sensors, Avantor®
Débitmètres
Measures low liquid flow rates with high accuracy.
  • Low cost plus high accuracy with ±3% of reading
  • Lightweight plastic design enables mounting in any position
These hall effect turbine flow rate sensors are ideal for OEM applications involving low flow liquid monitoring. The low cost coupled with 1/2% repeatability makes it an ideal candidate for replacing dispensing timer systems. Unlike existing timing systems, turbine technology is not influenced by changes in system pressure caused by aging filters. The sensor's standard power and output specifications make it easy to retrofit to existing controllers.