Imprimer
Masterflex® Adapter Fittings, Hose Barb to Male UNF Threaded, Tee, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX31209-50EA 61.9 EUR
MFLX31209-50 MFLX31209-51
Masterflex® Adapter Fittings, Hose Barb to Male UNF Threaded, Tee, Avantor®
Raccords et connecteurs
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Stainless Steel has very good chemical resistance
  • Temperature range of –40 to +200 °F (–40 to +93 °C)
  • Sterilize by ethylene oxide, autoclave, or gamma irradiation
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).