Imprimer
Masterflex® Adapter Fittings, Sanitary Ladish Clamp to Female Threaded, Straight, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40619-59
MFLX31806-03EA 244 EUR
MFLX31806-03 MFLX40619-60 MFLX40619-62 MFLX40619-67 MFLX40619-59
Masterflex® Adapter Fittings, Sanitary Ladish Clamp to Female Threaded, Straight, Avantor®
Fittings and Connectors
Maintain sanitary conditions in your processes.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
The sanitary clamp fittings may be autoclaved. The PVDF material tolerates higher temperatures better than polypropylene and performs more consistently after repeated autoclaving.