Imprimer
CPC® APC Acetal Quick-Disconnect Fittings, Bodies and Inserts
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06353-25BLK 06353-07BLK 06353-08BLK 06353-21BLK 06353-34BLK 06353-01BLK
MFLX06353-25BLKEA 220 EUR
MFLX06353-25BLK MFLX06353-07BLK MFLX06353-08BLK MFLX06353-21BLK MFLX06353-34BLK MFLX06353-01BLK
CPC® APC Acetal Quick-Disconnect Fittings, Bodies and Inserts
Raccords et connecteurs
Fittings connect in a snap.
  • No exposed metal parts
  • Protects against accidental disconnects
These CPC® APC-Series quick-disconnect fittings, combine a compact design with durable acetal construction, making them an excellent choice for many applications. All fittings feature a 1/4” flow, plastic thumb latch, fewer moving parts, and a smooth contoured design to deliver ease-of-use and excellent flow in a compact size. Simply press the thumb latch to disconnect fitting halves. Choose from in-line compression and hose barb bodies which feature shrouded latches to provide further protection against accidental disconnect. The ferruleless PTF compression fittings do not require ferrules to achieve a secure connection, which makes them easier to use and reuse. PTF fittings are designed for semi-rigid tubing, i.e., polyethylene, nylon, polyurethane, etc.
A complete coupler requires a fitting body and an insert (order separately).