Imprimer
Parker Hannifin Panel-Mount Needle Valves, PFA
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
98402-01 98402-02 06373-23 98402-03
MFLX98402-01EA 222 EUR
MFLX98402-01 MFLX98402-02 MFLX06373-23 MFLX98402-03
Parker Hannifin Panel-Mount Needle Valves, PFA
Vannes Needle Valves
Easily control flow with PFA needle valves.
  • Constructed of chemically inert PFA
When used with fluoropolymer tubing, these valves complete a chemically inert fluid pathway.