Imprimer
Warren Rupp Surge Suppressors and Dampeners
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
75008-00 75008-20 75008-30
MFLX75008-00EA 1010 EUR
MFLX75008-00 MFLX75008-20 MFLX75008-30
Warren Rupp Surge Suppressors and Dampeners
Pompe Pompes à membrane
Minimize pulsation of positive displacement pumps.
  • Less vibration and noise
  • Simple installation
  • Variety of materials of construction to meet the system conditions
These Warren Rupp surge suppressors and dampeners are ideal for use to minimize pulsation while pumping with your positive displacement pump. Choose from a wide range of materials based on the chemical compatibility required.