Imprimer
Micropump Pump Head Adapter Kits
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07003-99 07003-96
MFLX07104-48EA 677 EUR
MFLX07104-48 MFLX07003-99 MFLX07003-96
Micropump Pump Head Adapter Kits
Pompe Gear Pumps
Easily adapt your pump head to your drive with these adapter kits.
  • Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.