Imprimer
Lever-Action Drum Pumps with Nonmetallic-Body
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06511-50 06511-60 06511-80
MFLX06511-50EA 59.3 EUR
MFLX06511-50 MFLX06511-60 MFLX06511-80
Lever-Action Drum Pumps with Nonmetallic-Body
Pompe Pompes pour conteneur/transfert
Economical pumps for use in cabinets or tight work spaces.
  • Fit 15, 30, and 55-gallon drums with 2" IPS bung openings
  • Designed for use in industrial, agricultural, labs, and automotive industries
Choose from center-level pump and lever-action pump model options.
Center-Lever Pump is self-priming and features an anti-drip spout to eliminate wasted fluid. This pump is ideal for oils, detergents, and other compatible industrial fluids.
Lever-Action Pumps feature a curved discharge spout for convenient hose extension. Choose from three models depending on your chemical needs. Model 06511-50 is ideal for water, alcohol, gasoline, diesel, kerosene, and lubricants. Model 06511-60 handles the same fluids, plus water-based chemicals, acids, and alkalis. Model 06511-80 handles the same fluids, plus strong acids and strong alkalis.