Imprimer
Masterflex® Solenoid-Operated Pinch Valve For Masterflex® Pump Tubing, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
98306-02 98306-04 98306-08 98306-16 98306-18 98306-22 98306-26 98306-00 98306-10 98306-12 98306-14 98306-20 98306-24 98306-28 98306-30
MFLX98306-02EA 196 EUR
MFLX98306-02 MFLX98306-04 MFLX98306-08 MFLX98306-16 MFLX98306-18 MFLX98306-22 MFLX98306-26 MFLX98306-00 MFLX98306-10 MFLX98306-12 MFLX98306-14 MFLX98306-20 MFLX98306-24 MFLX98306-28 MFLX98306-30
Masterflex® Solenoid-Operated Pinch Valve For Masterflex® Pump Tubing, Avantor®
Vannes Solenoid Valves
Specifically designed for Masterflex® pump tubing.
  • Use with tubing that has durometer rating of 50 or less such as C-Flex® or silicone—7.25 psi max
  • Excellent for clean processes—fluid stays contained within tubing
  • Use for on/off flow control applications
Easily control the flow through your Masterflex tubing.