Imprimer
Black Rubber Vacuum Transfer Pump Tubing
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
96143-01 96143-02 96143-04 96143-06 96143-08 96143-10 96143-12 96143-14 96143-16
MFLX96143-01EA 133 EUR
MFLX96143-01 MFLX96143-02 MFLX96143-04 MFLX96143-06 MFLX96143-08 MFLX96143-10 MFLX96143-12 MFLX96143-14 MFLX96143-16
Black Rubber Vacuum Transfer Pump Tubing
Tuyaux Transfer Tubing
Ideal for vacuum and pressure applications.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Vacuum and pressure applications for general lab and industrial use
Characteristics: Thick wall, flexible, opaque
Temperature Range: 0 °F to 150 °F
Sterilize: Ethylene oxide or autoclave for 30 minutes at 15 psi