Imprimer
Tris base buffer, powder, Corning®
Marques : Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
61-233-RM 61-233-RT 61-233-RR
MDTC61-233-RMEA 124 EUR
MDTC61-233-RM MDTC61-233-RT MDTC61-233-RR
Tris base buffer, powder, Corning®
Tampons Tampons d'électrophorèse Tampons de migration
Tris is commonly used in buffer solutions in biochemical and molecular biological research.
This primary amine has an effective pH range of 7,0 to 9,2 and is reported to inhibit a number of different enzymes, limiting its usefulness in protein studies to some extent. This tris base buffer is provided in powder form.