Imprimer
Medium 199 (Mod.), Corning®
Marques : Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
50-050-PB 90-050-PB
50-050-PBEA 53.8 EUR
50-050-PB 90-050-PB
Medium 199 (Mod.), Corning®
Milieux de culture cellulaire
Medium 199 is often used in vaccine production, virology, and for the culture of various non-transformed cells
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It was originally developed as the result of efforts to create a fully defined, animal origin-free medium. Long-term use of this medium requires serum supplementation.