Imprimer
Dow Corning® Pharma-50 Transfer Tubing
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX96105-00EA 136 EUR
MFLX96105-00 MFLX96105-03 MFLX96105-04 MFLX96105-05 MFLX96105-08 MFLX96105-12 MFLX96105-02 MFLX96105-09 MFLX96105-11
Dow Corning® Pharma-50 Transfer Tubing
Tuyaux Transfer Tubing
Pharma tubing remains flexible and resilient for long service life.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotechnology , and ultra-pure fluid processing.
Characteristics: 50 durometer, translucent platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Pharma tubing remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silica to silicone tubing.
Certifications: Elastomer exceeds test guidelines for USP Class VI Plastics, and the tubing itself is FDA compliant, and meets 3A Sanitary Standards and European Pharmacopoeia (EP) requirements. Produced under Bulk Pharmaceutical (BP) and current GMP principles.
Temperature range: –60 to 460 °F (–51 to 232 °C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation.