Imprimer
CPC® ChemQuik® Quick-Disconnect Fittings, In-Line Coupling Bodies
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
31603-01 31603-15BLK
MFLX31603-02EA 238 EUR
MFLX31603-02 MFLX31603-01 MFLX31603-15BLK
CPC® ChemQuik® Quick-Disconnect Fittings, In-Line Coupling Bodies
Raccords et connecteurs
All plastic construction for high-purity use.
  • 100% metal-free couplings are ideal for ultrapure fluids, deionized water, and aggressive chemical applications
These ChemQuik® quick-disconnect couplings provide easy single-handed disconnection. These couplings feature valves to provide flow shut-off. The all plastic construction and lubricant-free design enables use in the most demanding applications.
For a complete coupling, order one body and one insert of the same size.