Imprimer
Centrifugal Multi-Purpose Separator/Extractor Weirs
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17315-20 17315-21 17315-22 17315-23 17315-24
MFLX17315-20EA 40.2 EUR
MFLX17315-20 MFLX17315-21 MFLX17315-22 MFLX17315-23 MFLX17315-24
Centrifugal Multi-Purpose Separator/Extractor Weirs
Pompe Centrifugal Separators
Mix, react, and/or separate liquid-liquid mixtures in a single device.
  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.