Imprimer
John Guest® Speedfit Ball Valves, Polypropylene
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
07391-00 07391-02 07391-04 07391-06 07391-08 07391-10
MFLX07391-00EA 19.4 EUR
MFLX07391-00 MFLX07391-02 MFLX07391-04 MFLX07391-06 MFLX07391-08 MFLX07391-10
John Guest® Speedfit Ball Valves, Polypropylene
Vannes Ball Valves
Quickly and easily connect and disconnect valve.
  • Constructed of durable polypropylene (PP)
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.