Imprimer
Masterflex® Flow-Through Pump Tubing Weights, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX07570-04EA 158 EUR
MFLX07570-04 MFLX78226-81 MFLX78226-82
Masterflex® Flow-Through Pump Tubing Weights, Avantor®
Weights Sinkers
Keep your samples flowing and tubing from curling.
  • Choose between PTFE or stainless steel construction
This flow-through tubing weight prevents tubing from curling or floating. Ideal for groundwater sampling, this weight fits Masterflex®; L/S® 15 or L/S® 24 Precision tubing.