Imprimer
Masterflex® PTFE Ball Valves, Female NPT, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
98514-01 98514-05 98514-09
MFLX98514-01EA 1290 EUR
MFLX98514-01 MFLX98514-05 MFLX98514-09
Masterflex® PTFE Ball Valves, Female NPT, Avantor®
Vannes Ball Valves
PTFE wetted parts allow for maximum chemical resistance.
  • Bidirectional 1/4-turn from OFF to ON
  • PTFE and polypropylene (PP) body construction
  • Perfect for use in harsh environments
These Masterflex® PTFE ball valves are ideal for use in semiconductor or pharmaceutical manufacturing and chemical processing.