Imprimer
Plug Valves with Compression Fittings, PFA
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
98402-05 98402-06
MFLX98402-05EA 300 EUR
MFLX98402-05 MFLX98402-06
Plug Valves with Compression Fittings, PFA
Vannes Plug Valves
Chemically-inert PFA handles most aggressive chemicals.
  • Constructed of durable PFA
Plug valves are FDA-approved and feature PFA seals and nuts.