Imprimer
Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX06832-01EA 45.5 EUR
MFLX06832-01 MFLX06832-02 MFLX06832-03 MFLX06832-04 MFLX06832-06 MFLX06832-08 MFLX06832-10 MFLX06832-12 MFLX06832-14 MFLX06832-18 MFLX06832-20 MFLX06832-22 MFLX06832-24 MFLX06832-28 MFLX06832-32 MFLX95613-00 MFLX95614-00 MFLX95614-01
Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Pinces Clips pour tubes
Reusable white acetal copolymer clamps will not rust or corrode.
  • Resists corrosion
Recommended for continuous use from ambient temperature to 200ºF (93ºC). Clamps are nonconducting and stable in a wide variety of chemicals.
Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers.