Imprimer
Masterflex® Plastic Tubing Clamp Ties, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX95615-00EA 44.5 EUR
MFLX95615-00 MFLX95615-10
Masterflex® Plastic Tubing Clamp Ties, Avantor®
Liens
Special curved clamp tie head ensures tight seal.
  • Ideal for general-purpose, single-use applications
Perfect for flexible fuel line, water line, and vacuum line applications. Ties measure 8" L x 0.18" W; maximum diameter of 1-3/4".