Imprimer
Masterflex® Transfer Tubing, Fluorinated Etyhlene Propylene (FEP), Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06450-01 06450-14
MFLX06406-12EA 101 EUR
MFLX06406-12 MFLX06406-18 MFLX06406-60 MFLX06406-62 MFLX06406-63 MFLX06406-64 MFLX06406-66 MFLX06406-72 MFLX06406-74 MFLX06406-76 MFLX06450-00 MFLX06450-02 MFLX06450-04 MFLX06450-05 MFLX06450-06 MFLX06450-07 MFLX06450-08 MFLX06450-09 MFLX06450-15 MFLX06450-17 MFLX06450-19 MFLX06450-01 MFLX06450-14
Masterflex® Transfer Tubing, Fluorinated Etyhlene Propylene (FEP), Avantor®
Tuyaux Transfer Tubing
Does not deteriorate with age.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work.
Characteristics: Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesn’t deteriorate with age. Tough, but bends even at -454 °F (-270 °C). Translucent.
Certifications: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured in ISO 9001-2008 approved facility.
Temperature range: -454 to 400 °F (-270 to 205 °C)
Sterilize: Autoclave, ethylene oxide, or use dry heat.