Imprimer
Masterflex® Transfer Tubing, Peroxide-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX06411-03EA 161 EUR
MFLX06411-03 MFLX06411-19 MFLX06411-55 MFLX06411-58 MFLX06411-59 MFLX06411-60 MFLX06411-62 MFLX06411-63 MFLX06411-64 MFLX06411-65 MFLX06411-66 MFLX06411-67 MFLX06411-68 MFLX06411-69 MFLX06411-70 MFLX06411-71 MFLX06411-72 MFLX06411-74 MFLX06411-75 MFLX06411-76 MFLX06411-77 MFLX06411-78 MFLX06411-79 MFLX06411-80 MFLX06411-82 MFLX06411-83 MFLX06411-12
Masterflex® Transfer Tubing, Peroxide-Cured Silicone, Avantor®
Tuyaux Transfer Tubing
No need to worry about the heat - use for applications up to 460 °F.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Pharmaceutical, biotechnology, food and beverage, aerospace, and general lab applications
Characteristics: Peroxide-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent
Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -60 to 460 °F (-51 to 238 °C)
Sterilize: Autoclave or gamma irradiation