Imprimer
Hand-Operated Piston Dispensing Drum Pump, Polypropylene
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06512-00 06512-05
MFLX06512-00EA 25.6 EUR
MFLX06512-00 MFLX06512-05
Hand-Operated Piston Dispensing Drum Pump, Polypropylene
Pompe Pompes pour conteneur/transfert
Economical piston pumps are ideal for a variety of fluids.
  • Adaptable to most pails and drums
Models 06512-00 and -05 have adjustable strokes to allow for dispensing different amounts fo fluid with just one pump. Model 06512-00 pumps 64, 32, 22 and 16 strokes per gallon, model 60512-05 pumps 32, 16, 11 and 8 strokes per gallon.