Imprimer
Masterflex® L/S® Replacement Brushes and Brush Caps, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX07520-04EA 64.2 EUR
MFLX07520-04 MFLX07520-06 MFLX07533-01
Masterflex® L/S® Replacement Brushes and Brush Caps, Avantor®
Pompe Pompes péristaltiques
Quickly and easily replace components of your Masterflex system.
  • Choose replacement brushes and caps based on your Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.